missing translation for 'onlineSavingsMsg'
Learn More

Chloropretadalafil-13C2, TRC

Product Code. 30366627
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30366627 2.5 mg 2.5mg
30365020 10 mg 10mg
30363475 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30366627 Supplier Toronto Research Chemicals Supplier No. TRCC3813642.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula 13C2 C20 H19 Cl N2 O5
Molecular Weight (g/mol) 428.835
Synonym (1R,3R)-1-(1,3-Benzodioxol-5-yl)-2-(2-chloroacetyl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylic-13C2 Acid Methyl Ester, (1R,3R)- 1-(1,3-Benzodioxol-5-yl)-2-(chloroacetyl)-2,3,4,9-tetrahydro-1H-Pyrido[3,4-b]indole-3-carboxylic-13C2 Acid Methyl Ester, (1R-cis)-1-(1,3-Benzodioxol-5-yl)-2-(chloroacetyl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylic-13C2 Acid Methyl Ester, (1R,3R)-1-(1,3-Benzodioxol-5-yl)-2-(chloroacetyl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylic-13C2 Acid Methyl Ester, Methyl (1R,3R)-1-(1,3-Benzodioxol-5-yl)-2-(chloroacetyl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylate-13C2
IUPAC Name methyl (1R,3R)-1-(1,3-benzodioxol-5-yl)-2-(2-chloroacetyl)-1,3,4,9-tetrahydropyrido[3,4-b]indole-3-carboxylate
SMILES COC(=O)[C@H]1Cc2c([nH]c3ccccc23)[C@H](N1[13C](=O)[13CH2]Cl)c4ccc5OCOc5c4

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C22H19ClN2O5/c1-28-22(27)16-9-14-13-4-2-3-5-15(13)24-20(14)21(25(16)19(26)10-23)12-6-7-17-18(8-12)30-11-29-17/h2-8,16,21,24H,9-11H2,1H3/t16-,21-/m1/s1/i10+1,19+1
Formula Weight 428.105
Chemical Name or Material Chloropretadalafil-13C2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.