missing translation for 'onlineSavingsMsg'
Learn More

Chlorotri(1-pyrrolidinyl)phosphonium hexafluorophosphate, 98+%, Thermo Scientific™

Product Code. 15421496
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15421496

Brand: Thermo Scientific Alfa Aesar H54693.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133894-48-1
Molecular Formula C12H24ClF6N3P2
Molecular Weight (g/mol) 421.733
MDL Number MFCD00210035
InChI Key BSCYRXJVGSZNKX-UHFFFAOYSA-N
Synonym pyclop, chlorotripyrrolidinophosphonium hexafluorophosphate, chlorotripyrrolidinophosphonium hexa-fluorophosphate, chlorotri 1-pyrrolidinyl phosphonium hexafluorophosphate, 98+%, pyciop, ref dupl: pyclop, chlorotripyrrolidino, acmc-1c01w, ksc177a1n, chlorotripyrrolidinophosphoniumhexafluorophosphat
PubChem CID 16211753
IUPAC Name chloro(tripyrrolidin-1-yl)phosphanium;hexafluorophosphate
SMILES C1CCN(C1)[P+](N2CCCC2)(N3CCCC3)Cl.F[P-](F)(F)(F)(F)F

Specifications

Melting Point 145°C
Quantity 1 g
UN Number UN1759
Beilstein 6842341
Sensitivity Moisture sensitive
Formula Weight 421.73
Percent Purity ≥98%
Chemical Name or Material Chlorotri(1-pyrrolidinyl)phosphonium hexafluorophosphate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.