missing translation for 'onlineSavingsMsg'
Learn More

Cholesterol, Extra Pure, SLR, Fisher Chemical

Product Code. 10263660 Shop All Fisher Chemical Products
Change view
Click to view available options
Packaging:
Plastic powder jar
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Unit Size
10263660 Plastic powder jar 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10263660 Supplier Fisher Chemical Supplier No. C/5360/48

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 57-88-5
Melting Point 147°C
Color White
Packaging Plastic powder jar
Molecular Formula C27H46O
MDL Number 3646
Formula Weight 386.65g/mol
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.