missing translation for 'onlineSavingsMsg'
Learn More

Cholesteryl N-Butyrate, TRC

Product Code. 30133910
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133910 50 g 50g
30137041 10 g 10g
30010981 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133910 Supplier Toronto Research Chemicals Supplier No. TRCC43263350G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 521-13-1
Molecular Formula C31H52O2
Molecular Weight (g/mol) 456.74
IUPAC Name [(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] butanoate
SMILES CCCC(O[C@H]1CC[C@]2(C(C1)=CC[C@H]3[C@@H]4CC[C@@H]([C@]4(CC[C@@H]32)C)[C@@H](CCCC(C)C)C)C)=O

Specifications

Quantity 50 g
InChI Formula InChI=1S/C31H52O2/c1-7-9-29(32)33-24-16-18-30(5)23(20-24)12-13-25-27-15-14-26(22(4)11-8-10-21(2)3)31(27,6)19-17-28(25)30/h12,21-22,24-28H,7-11,13-20H2,1-6H3/t22-,24+,25+,26-,27+,28+,30+,31-/m1/s1
Formula Weight 456.3967
Chemical Name or Material Cholesteryl N-Butyrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.