missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHRNA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10613-1-AP-20UL
This item is not returnable.
View return policy
Description
Acetylcholine receptors (AChRs) mediate synaptic transmission at the neuromuscular junction. The channel-linked AChR that mediates rapid, excitatory actions of acetylcholine is called nicotinic AChR (nAChR) because it can be activated by nicotine. The non-channel linked AChR that medicates the slow actions of acetylcholine, which can be either inhibitory or excitatory, is called muscarinic AChR (mAChR) because it can be activated by muscarine. The nAChRs fall into two subclasses, the muscle-type and the neuronal-type. The muscle-type receptor is a pentameric glycoprotein of five membrane spanning subunits, (2 alpha 1′s, beta 1, gamma or epsilon and delta) which form a ligand gated ion channel. The neuronal nAChR is also a pentamer, but unlike the muscle-type, is made up of only two general types of subunits, alpha and beta of which multiple subtypes have been described (alpha 2 to alpha 8 and beta 2 to beta 4).Specifications
| CHRNA1 | |
| Polyclonal | |
| Unconjugated | |
| CHRNA1 | |
| ACHRA, ACHRD, CHNRA, CHRNA, CHRNA1, CMS2A, FCCMS, nAChRA1, SCCMS | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1134, 11435, 79557 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P02708, P04756, P25108 | |
| Chrna1 | |
| CHRNA1 Fusion Protein Ag0933 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction