missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHURC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12247-1-AP-20UL
This item is not returnable.
View return policy
Description
The MW of this protein is 13 kDa, and 12247-1-AP specifically recognizes the 13 kDa protein.
Transcriptional activator that mediates FGF signaling during neural development. Plays a role in the regulation of cell movement. Does not bind DNA by itself.Specifications
| CHURC1 | |
| Polyclonal | |
| Unconjugated | |
| CHURC1 | |
| C14orf52, chch, CHURC1, churchill domain containing 1, My015, Protein Churchill | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 211151, 299154, 91612 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6DG52, Q8WUH1 | |
| CHURC1 | |
| CHURC1 Fusion Protein Ag2892 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction