missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHURC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | CHURC1 |
|---|---|
| Concentration | 0.15 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16899843
|
Proteintech
12247-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16889843
|
Proteintech
12247-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The MW of this protein is 13 kDa, and 12247-1-AP specifically recognizes the 13 kDa protein.
Transcriptional activator that mediates FGF signaling during neural development. Plays a role in the regulation of cell movement. Does not bind DNA by itself.Specifications
| CHURC1 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Rat, Human, Mouse | |
| Q6DG52, Q8WUH1 | |
| 211151, 299154, 91612 | |
| CHURC1 Fusion Protein Ag2892 | |
| Primary | |
| -20°C | |
| CHURC1 |
| 0.15 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| C14orf52, chch, CHURC1, churchill domain containing 1, My015, Protein Churchill | |
| CHURC1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title