missing translation for 'onlineSavingsMsg'
Learn More

Cilnidipine-d3, TRC

Product Code. 30352072
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30352072 5 mg 5mg
30360909 10 mg 10mg
30366282 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30352072 Supplier Toronto Research Chemicals Supplier No. TRCC4413775MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C27 D3 H25 N2 O7
Molecular Weight (g/mol) 495.539
Synonym 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid 2-Methoxyethyl-d3 (2E)-3-Phenyl-2-propenyl Ester, FRC-8653-d3, Atelec-d3, Cinalong-d3, Siscard-d3
IUPAC Name 5-O-[(E)-3-phenylprop-2-enyl] 3-O-[2-(trideuteriomethoxy)ethyl] 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
SMILES [2H]C([2H])([2H])OCCOC(=O)C1=C(C)NC(=C(C1c2cccc(c2)[N+](=O)[O-])C(=O)OC\C=C\c3ccccc3)C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C27H28N2O7/c1-18-23(26(30)35-14-8-11-20-9-5-4-6-10-20)25(21-12-7-13-22(17-21)29(32)33)24(19(2)28-18)27(31)36-16-15-34-3/h4-13,17,25,28H,14-16H2,1-3H3/b11-8+/i3D3
Formula Weight 495.208
Chemical Name or Material Cilnidipine-d3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.