missing translation for 'onlineSavingsMsg'
Learn More

CIZ1 Antibody, FITC, Novus Biologicals™

Product Code. 18542801 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18542801 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18542801 Supplier Novus Biologicals Supplier No. NB10074624F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CIZ1 Polyclonal antibody specifically detects CIZ1 in Human, Mouse samples. It is validated for Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen CIZ1
Applications Immunofluorescence
Classification Polyclonal
Conjugate FITC
Dilution Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias CDKN1A interacting zinc finger protein 1, CDKN1A-interacting zinc finger protein 1, cip1-interacting zinc finger protein, LSFR1ZNF356Nuclear protein NP94, NP94, Zinc finger protein 356
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 848 and 898 of human CDKN1A interacting zinc finger protein 1 using the numbering given in entry NP_036259.2 (GeneID 25792).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 25792
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.