missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CL 82198 hydrochloride
GREENER_CHOICE

Product Code. 18034762
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18034762 10 mg 10mg
18045502 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18034762 Supplier Tocris Bioscience™ Supplier No. 2632/10
  • ONLINE EXCLUSIVE PRICE
    161.00€
    152.25€ / 10mg
    Save 8.75€
    5% Off
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Selective inhibitor of MMP-13

Selective inhibitor of MMP-13 (89% inhibition at 10 μg/mL) that displays no activity at MMP-1, MMP-9 or TACE. Inhibits in vitro invasion by the human pituitary adenoma cell line HP75. Rescues paclitaxel induced axon degradation and reduces associated neurotoxicity in zebrafish.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1188890-36-9
Molecular Formula C17H23ClN2O3

Specifications

Quantity 10 mg
Formula Weight 338.83
Purity >98%
Molecular Weight (g/mol) 338.832
Chemical Name or Material CL 82198 hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.