missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CLK4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15708331 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15708331 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15708331 Supplier Invitrogen™ Supplier No. PA560494

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: EYRNDYCEGY VPRHYHRDIE SGYRIHCSKS SVRSRRSSPK RKRNRHCSSH QSRSKSHR Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 52%.

CLK4 belongs to the CDC2-like protein kinase (CLK) family. This protein kinase can interact with and phosphorylate the serine- and arginine-rich proteins, which are known to play an important role in the formation of spliceosomes, and thus may be involved in the regulation of alternative splicing. Studies in the Israeli sand rat Psammomys obesus suggested that the ubiquitin-like 5, a highly conserved ubiquitin-like protein, may interact with and regulate the activity of this kinase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLK4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CLK4
Gene Accession No. Q9HAZ1
Gene Alias AI987988; C85119; CDC like kinase 4; CDC-like kinase 4; Clk4; DKFZp686A20267; Dual specificity protein kinase CLK4; protein serine threonine kinase Clk4
Gene Symbols CLK4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CLK4. Recombinant protein control fragment (Product #RP-100448).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57396
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.