missing translation for 'onlineSavingsMsg'
Learn More

CLN6 Polyclonal Antibody, Invitrogen™

Product Code. 15507681
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15507681 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15507681 Supplier Invitrogen Supplier No. PA536442

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 40 kDa. Purity is >95% by SDS-PAGE.

CLN6 Polyclonal antibody specifically detects CLN6 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLN6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene CLN6
Gene Accession No. Q9NWW5
Gene Alias 1810065L06Rik; AW743417; ceroid-lipofuscinosis; ceroid-lipofuscinosis neuronal 6 late infantile; ceroid-lipofuscinosis neuronal protein 6; ceroid-lipofuscinosis, neuronal 6; ceroid-lipofuscinosis, neuronal 6, late infantile, variant; CLN4A; CLN6; D9Bwg1455e; HsT18960; nclf; neuronal ceroid lipofuscinosis; Protein CLN6
Gene Symbols CLN6
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 229-274 of human CLN6
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 315746, 54982, 76524
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.