missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CNIH1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15817833
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15817833 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15817833 Supplier Invitrogen™ Supplier No. PA569420

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 79%; Rat: 100%; Zebrafish: 100%.

CNIH is involved in the selective transport and maturation of TGF-alpha family proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CNIH1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CNIH1
Gene Accession No. O35372
Gene Alias 0610007J15; Cnih; CNIH1; CNIH-1; CNIL; Cornichon family AMPA receptor auxiliary protein 1; cornichon homolog; cornichon homolog 1 (Drosophila); cornichon-like isoform 1; hypothetical protein LOC613143; protein cornichon homolog; Protein cornichon homolog 1; T-cell growth-associated molecule 77; TGAM77; UNQ155/PRO181; wu:fc21b06; wu:fi12c05; zgc:112228; zgc:56103
Gene Symbols CNIH1
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of rat CNIH1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12793, 289994
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.