missing translation for 'onlineSavingsMsg'
Learn More

Coactosin-like Protein 1/CotL1 Antibody (1D6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-31203-0.05ML

 View more versions of this product

Product Code. 18161815

  • 362.00€ / 0.05mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Coactosin-like Protein 1/CotL1 Monoclonal specifically detects Coactosin-like Protein 1/CotL1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Coactosin-like Protein 1/CotL1
Monoclonal
1 mg/mL
Western Blot 1:1000, ELISA 1:1000
CLPFLJ43657, coactosin-like 1 (Dictyostelium), coactosin-like protein, MGC19733
Mouse
Protein A purified
RUO
23406
Protein A purified
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG2b κ
Western Blot, ELISA
1D6
Unconjugated
PBS (pH 7.4) and 10% Glycerol with 0.02% Sodium Azide
COTL1
Recombinant human COTL1 (1-142aa) purified from E. coli
0.05 mL
Primary
Anti-human COTL1 mAb, clone 1D6, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human COTL1 protein.
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only