missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coactosin-like Protein 1/CotL1 Antibody (1D6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
362.00€ - 488.00€
Specifications
| Antigen | Coactosin-like Protein 1/CotL1 |
|---|---|
| Clone | 1D6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, ELISA 1:1000 |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18161815
|
Novus Biologicals
NBP2-31203-0.05ML |
0.05 mL |
362.00€
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18175725
|
Novus Biologicals
NBP2-31203-0.1ML |
0.1 mL |
488.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Coactosin-like Protein 1/CotL1 Monoclonal specifically detects Coactosin-like Protein 1/CotL1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
| Coactosin-like Protein 1/CotL1 | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| CLPFLJ43657, coactosin-like 1 (Dictyostelium), coactosin-like protein, MGC19733 | |
| COTL1 | |
| IgG2b κ | |
| Protein A purified | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1D6 | |
| Western Blot 1:1000, ELISA 1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.4) and 10% Glycerol with 0.02% Sodium Azide | |
| 23406 | |
| Recombinant human COTL1 (1-142aa) purified from E. coli | |
| Primary | |
| Protein A purified | |
| Anti-human COTL1 mAb, clone 1D6, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human COTL1 protein. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title