missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Connexin 37 Polyclonal Antibody

Product Code. 10241513
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10241513 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10241513 Supplier Invitrogen™ Supplier No. 404200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

40-4200 was used in the immunofluorescence (IHC) analysis to successfully detect Connexin 37 in mouse heart sections.

This gene encodes a member of the connexin gene family. The encoded protein is a component of gap junctions, which are composed of arrays of intercellular channels that provide a route for the diffusion of low molecular weight materials from cell to cell. Mutations in this gene have been associated with atherosclerosis and a higher risk of myocardial infarction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Connexin 37
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene GJA4
Gene Accession No. P28235
Gene Alias AU020209; AW558810; Cnx37; connexin 37; connexin-37; CX37; CXN37; Cxn-37; gap junction alpha-4 protein; gap junction membrane channel protein alpha 4; Gap junction membrane channel, protein alpha 4 (connexin 37); gap junction protein alpha 4; gap junction protein, alpha 4; gap junction protein, alpha 4, 37kDa; GJA4; Gja-4
Gene Symbols GJA4
Host Species Rabbit
Immunogen Synthetic peptide derived from the C-terminal region of the mouse connexin 37 protein.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14612
Target Species Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.