Learn More
Corning™ Primaria™ Tissue Culture Flasks
GREENER_CHOICE
Product Code. 10196070 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
50 mL
250 mL
Culture Area:
25 cm2
75 cm2
Closure Type:
Plug Seal
Vented
Neck Style:
Canted
Straight
Unit Size:
Pack of 100
Pack of 200
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Culture Area Closure Type Neck Style unitSize
10196070 250 mL 75 cm2 Plug Seal Straight Pack of 100
10613672 50 mL 25 cm2 Vented Canted Pack of 100
10469222 250 mL 75 cm2 Vented Straight Pack of 100
10556932 50 mL 25 cm2 Plug Seal Canted Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10196070 Supplier Corning™ Supplier No. 353824
Estimated Shipment: 08-12-2026
Add to basket
Add to basket
Improves attachment, spreading and growth for many primary cells or cell lines. Corning™ Primaria™ Tissue Culture Flasks are suitable for growing cells that grow poorly on conventional, negatively charged, standard tissue culture surfaces—including tumor cells, endothelial cells and hepatocytes.
  • Unique nitrogen-containing tissue culture surface chemistry
  • Improves attachment, spreading, and growth for many primary cells or cell lines
  • Crystal-grade polystyrene modified by proprietary vacuum-gas plasma treatment process
  • Stable, permanent surface modification
  • Optically clear
  • No special storage required
  • Samples from each lot of Corning Primaria products are analyzed by Electron Scanning for Chemical Analysis (ESCA)
  • Packaged in red color-coded, peel-open, medical-style packages
  • Sterilized by gamma irradiation
  • Nonpyrogenic tested to less than 0.1 EU/mL
Order Info
Smaller flasks packed 20 per sleeve in cases of 200 (08-772-45 in cases of 100); larger flasks packed five per sleeve in cases of 100
TRUSTED_SUSTAINABILITY
Specifications
Capacity (English) 8.45 oz.
Closure Color Blue
Closure Material Phenolic
Color Optically Clear
Capacity (Metric) 250 mL
Material Polystyrene
For Use With (Application) Cell culture, tissue culture, cell growth, tumor cell culture, endothelial cell culture, hepatocyte culture
Length (English) 5.93 in.
Neck Style Straight
No. per Case 100/Cs.
No. per Pack 5/Pk.
Product Type Cell Culture Flask
Shape Rectangular
Temperature Range (Metric) 4°C to 60°C
Width (English) 3.19 in.
Autoclavable Not Autoclavable
Disposable Single-use
Graduated Yes
Sterility Sterile
Bottom Shape Rectangular
Closure Type Plug Seal
Culture Area 75 cm2
Volume (Metric) Working 15 to 22.5 mL
Closure Size 22 mm
Quantity 100/Cs.
Dimensions (L x W x H) 150.7 x 80.9 x 38.1 mm
Show More Show Less
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.