missing translation for 'onlineSavingsMsg'
Learn More

COSMC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16898934 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16898934 20 μL 20µL
16888934 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16898934 Supplier Proteintech Supplier No. 192541AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes a type II transmembrane protein that is similar to the core 1 beta1,3-galactosyltransferase 1, which catalyzes the synthesis of the core-1 structure, also known as Thomsen-Friedenreich antigen, on O-linked glycans. This gene product lacks the galactosyltransferase activity itself, but instead acts as a molecular chaperone required for the folding, stability and full activity of the core 1 beta1,3-galactosyltransferase 1. Mutations in this gene have been associated with Tn syndrome. Alternatively spliced transcript variants encoding the same protein have been identified.

Specifications

Antigen COSMC
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene C1GALT1C1
Gene Accession No. Q499P3, Q96EU7, Q9JMG2
Gene Alias C1Gal T2, C1GALT1 chaperone 1, C1GALT1 specific chaperone 1, C1GALT1C1, C1GALT2, c38h2 l1, C38H2 like protein 1, Core 1 beta3 Gal T2, COSMC, HSPC067, MSTP143, UNQ273/PRO310
Gene Symbols C1galt1c1
Host Species Rabbit
Immunogen COSMC Fusion Protein Ag6899
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29071, 302499, 59048
Target Species Rat, Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.