missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CP 99994 dihydrochloride
GREENER_CHOICE

Product Code. 18043802
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18043802 50 mg 50mg
18004192 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18043802 Supplier Tocris Bioscience™ Supplier No. 3417/50
  • ONLINE EXCLUSIVE PRICE
    1310.00€
    1235.85€ / 50mg
    Save 74.15€
    5% Off
 Restricted Item
Estimated Shipment: 17-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High affinity NK1 antagonist

High affinity NK1 antagonist (Ki = 0.145 nM in vitro). Also displays high ex vivo binding potency in gerbil striatum (IC50 = 36.8 nM). Attenuates endothelium-dependent contraction induced by substance P.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145148-39-6
Molecular Formula C19H26Cl2N2O

Specifications

Quantity 50 mg
Formula Weight 369.33
Purity >99%
Molecular Weight (g/mol) 369.33
Chemical Name or Material CP 99994 dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.