Learn More
CPEB3 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12669-1-AP-150UL
 View more versions of this product
Product Code. 16820514
  • 410.00€ / 150µL
Description
The CPEB3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CPEB3. This antibody recognizes human, mouse, rat antigen. The CPEB3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
CPEB3
Polyclonal
Unconjugated
150 μL
22849
IgG
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
267 μg/mL
Rabbit
Primary
Human, Rat, Mouse