Learn More
CPEB3 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12669-1-AP-20UL
Description
The CPEB3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CPEB3. This antibody recognizes human, mouse, rat antigen. The CPEB3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| CPEB3 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 22849 | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 267 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |