missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CPI-17 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16350545
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16350545 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16350545 Supplier Invitrogen™ Supplier No. PA583590

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: VKYDRRELQR RLDVEKWIDG RLEELYRGME ADMPDEINID ELLELESEEE RSR Highest antigen sequence indentity to the following orthologs: Mouse - 89%, Rat - 92%.

CPI-17 is a phosphorylation-dependent inhibitory protein for smooth muscle myosin phosphate. CPI-17 was originally identified as a PKC-potentiated inhibitory protein of protein phosphatase-1, which is dominantly expressed in smooth muscle. Phosphorylation at Threonine 38, in vitro, by PKC or Rho-kinase enhances the inhibitory potency toward myosin phosphatase. CPI-17 is also phosphorylated at Threonine 38 by protein kinase N and might be involved in the calcium sensitization of smooth muscle contraction as a downstream effector of Rho and/or arachidonic acid. CPI-17 is dually phosphorylated at Serine 12 and Threonine 38 by a MYPT-associated kinase, M110 kinase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPI-17
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPP1R14A
Gene Accession No. Q96A00
Gene Alias 1110001M11Rik; 17 kDa PKC-potentiated inhibitory protein of PP1; 17-kDa PKC-potentiated inhibitory protein of PP1; 17-KDa protein; Cpi; CPI17; CPI-17; PKC-potentiated inhibitory protein of PP1; PPP1INL; PPP1R14A; Protein kinase C-potentiated inhibitor protein of 17 kDa; protein phosphatase 1 regulatory inhibitor subunit 14A; protein phosphatase 1 regulatory subunit 14A; protein phosphatase 1, regulatory (inhibitor) subunit 14A
Gene Symbols PPP1R14A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPP1R14A. Recombinant protein control fragment (Product #RP-102920).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94274
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.