missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CREB3L3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15791042 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15791042 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15791042 Supplier Invitrogen™ Supplier No. PA563270

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: DAVPGSEAPG PRPEADTTRE ESPGSPGADW GFQDTANLTN STEELDNATL VLRNATEGLG QVALLDWVAP GPSTGSGRAG LE Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 49%.

CREB3L3 is a liver-specific transcription factor and member of the CREB/ATF transcription factor family (Omori et al., 2001 [PubMed 11353085]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CREB3L3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CREB3L3
Gene Accession No. Q68CJ9
Gene Alias BC010786; cAMP responsive element binding protein 3 like 3; cAMP responsive element binding protein 3-like 3; cAMP-responsive element-binding protein 3-like protein 3; CREB/ATF family transcription factor; CREB3L3; CREBH; CREB-H; Cyclic AMP-responsive element-binding protein 3-like protein 3; D10Bur1e; HYST1481; Processed cyclic AMP-responsive element-binding protein 3-like protein 3; Transcription factor CREB-H
Gene Symbols CREB3L3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CREB3L3. Recombinant protein control fragment (Product #RP-103249).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84699
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.