missing translation for 'onlineSavingsMsg'
Learn More

CXCR5 Antibody (614641) [Alexa Fluor™ 532], Novus Biologicals™

Product Code. 18542526 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18542526 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18542526 Supplier Novus Biologicals Supplier No. FAB6198X

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

CXCR5 Monoclonal antibody specifically detects CXCR5 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-reported
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCR5
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 614641
Conjugate Alexa Fluor 532
Dilution Flow Cytometry, Immunocytochemistry, CyTOF-reported
Formulation 50mM Sodium Borate
Gene Alias BLR1MDR-15, Burkitt lymphoma receptor 1, Burkitt lymphoma receptor 1, GTP binding protein (chemokine (C-X-C motif)receptor 5), Burkitt lymphoma receptor 1, GTP-binding protein, CD185, CD185 antigen, chemokine (C-X-C motif) receptor 5, C-X-C chemokine receptor type 5, CXC-R5, MDR15CXCR-5, MGC117347, Monocyte-derived receptor 15
Host Species Rat
Immunogen E. coli-derived recombinant mouse CXCR5, Met1-Pro57, Accession # Q04683
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 643
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.