missing translation for 'onlineSavingsMsg'
Learn More

CYBB/NOX2 Antibody (NL7) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications

Brand:  Novus Biologicals NBP1-41012-0.025MG

 View more versions of this product

Product Code. 18080034

  • ONLINE EXCLUSIVE PRICE
    217.00€
    204.75€ / 25µg
    Save 12.25€
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CYBB/NOX2 Monoclonal specifically detects CYBB/NOX2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CYBB/NOX2
Monoclonal
1.0 mg/mL
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10-1:500
P04839
CYBB
Cytochrome B245 solubilised in octyl-beta-glucopyranoside
0.025 mg
Primary
This antibody recognises the heavy chain of cytochrome B245. It recognises the native as well as the denatured form of the protein
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1
Immunohistochemistry, Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, ELISA, Immunocytochemistry
NL7
Unconjugated
PBS (pH 7.4) with 0.09% Sodium Azide
CGD, CGD91-phox, Cytochrome b(558) subunit beta, cytochrome b-245 heavy chain, cytochrome b-245, beta polypeptide, Cytochrome b558 subunit beta, EC 1.6.3, GP91-1, GP91PHOX, GP91-PHOX, Heme-binding membrane glycoprotein gp91phox, NADPH oxidase 2, Neutrophil cytochrome b 91 kDa polypeptide, NOX2chronic granulomatous disease, p22 phagocyte B-cytochrome, p91-PHOX, Superoxide-generating NADPH oxidase heavy chain subunit
Mouse
Protein G purified
RUO
1536
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only