missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYBB/NOX2 Antibody (NL7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
217.00€ - 548.00€
Specifications
| Antigen | CYBB/NOX2 |
|---|---|
| Clone | NL7 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Immunohistochemistry, Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, ELISA, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18080034
|
Novus Biologicals
NBP1-41012-0.025MG |
0.025 mg |
217.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18776673
|
Novus Biologicals
NBP1-41012-0.1mg |
0.1 mg |
548.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CYBB/NOX2 Monoclonal specifically detects CYBB/NOX2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular).Specifications
| CYBB/NOX2 | |
| 1.0 mg/mL | |
| Immunohistochemistry, Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, ELISA, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P04839 | |
| 1536 | |
| Cytochrome B245 solubilised in octyl-beta-glucopyranoside | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| NL7 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.4) with 0.09% Sodium Azide | |
| CGD, CGD91-phox, Cytochrome b(558) subunit beta, cytochrome b-245 heavy chain, cytochrome b-245, beta polypeptide, Cytochrome b558 subunit beta, EC 1.6.3, GP91-1, GP91PHOX, GP91-PHOX, Heme-binding membrane glycoprotein gp91phox, NADPH oxidase 2, Neutrophil cytochrome b 91 kDa polypeptide, NOX2chronic granulomatous disease, p22 phagocyte B-cytochrome, p91-PHOX, Superoxide-generating NADPH oxidase heavy chain subunit | |
| CYBB | |
| IgG1 | |
| Protein G purified | |
| This antibody recognises the heavy chain of cytochrome B245. It recognises the native as well as the denatured form of the protein |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title