missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexan-d11-ol, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.50g
1g
Specifications
Specifications
| Chemical Name or Material | Cyclohexanol-d11 |
| CAS | 93131-17-0 |
| Molecular Formula | C6 D11 H O |
| Quantity | 1 g |
| Synonym | Cyclohexan-1,2,2,3,3,4,4,5,5,6,6-d11-ol (ACI), Cyclohexan-d11-ol (9 CI), Cyclohexanol-D11, Cyclohexanol-d11 |
| InChI Formula | InChI=1 S/C6H12O/c7-6-4-2-1-3-5-6/h6-7 H,1-5H2/i1D2,2D2,3D2,4D2,5D2,6 D |
| SMILES | [2 H]C1([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])(O)C([2 H])([2 H])C1([2 H])[2 H] |
| IUPAC Name | 1,2,2,3,3,4,4,5,5,6,6-undecadeuteriocyclohexan-1-ol |
| Molecular Weight (g/mol) | 111.2267 |
| Formula Weight | 111.1579 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction