missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, TRC

Product Code. 30012488
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Unit Size:
1 L
100mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012488 100 mL 100mL
30010204 500 mL 500mL
30005671 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012488 Supplier Toronto Research Chemicals Supplier No. TRCC991300100ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6 H12
Molecular Weight (g/mol) 84.16
Synonym Benzene, hexahydro-, Cactus Solvent CHX, Hexahydrobenzene, Hexamethylene, Hexanaphthene, NSC 406835, Ethanol Imp. E (EP)
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Quantity 100 mL
InChI Formula InChI=1S/C6H12/c1-2-4-6-5-3-1/h1-6H2
Formula Weight 84.0939
Chemical Name or Material Cyclohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.