missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylmethanol-d5 (Major), TRC

Product Code. 30364209
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30364209 250 mg 250mg
30352092 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30364209 Supplier Toronto Research Chemicals Supplier No. TRCC988607250MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclopropylmethanol-d5 (Major)
Molecular Formula C4 D5 H3 O
Quantity 250 mg
Synonym Cyclopropanemethanol-d5, (Hydroxymethyl)cyclopropane-d5, Cyclopropanemethyl-d5 Alcohol, Cyclopropylcarbinol-d5, Cyclopropylcarbinyl-d5 Alcohol, NSC 85925-d5
InChI Formula InChI=1S/C4H8O/c5-3-4-1-2-4/h4-5H,1-3H2/i1D2,2D2,4D
SMILES [2H]C1([2H])C([2H])([2H])C1([2H])CO
IUPAC Name (1,2,2,3,3-pentadeuteriocyclopropyl)methanol
Molecular Weight (g/mol) 77.136
Formula Weight 77.089
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.