Learn More
CYP20A1 Mouse anti-Human, Unconjugated, Clone: 2A9E2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67433-1-IG-20UL
 View more versions of this product
Product Code. 16820066
  • 145.00€ / 20µL
Description
The CYP20A1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CYP20A1. This antibody recognizes human, mouse, rat antigen. The CYP20A1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
CYP20A1
Monoclonal
500 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, ELISA
2A9E2
Unconjugated
20 μL
57404