Learn More
CYP20A1 Mouse anti-Human, Unconjugated, Clone: 2A9E2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67433-1-IG-20UL
Description
The CYP20A1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CYP20A1. This antibody recognizes human, mouse, rat antigen. The CYP20A1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| CYP20A1 | |
| Monoclonal | |
| 500 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunohistochemistry, ELISA | |
| 2A9E2 | |
| Unconjugated | |
| 20 μL | |
| 57404 | |
| IgG1 |