Learn More
CYP20A1 Mouse anti-Human, Unconjugated, Clone: 2A9E2, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | CYP20A1 |
|---|---|
| Clone | 2A9E2 |
| Concentration | 500 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Monoclonal |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
16820066
|
Proteintech
67433-1-IG-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16810066
|
Proteintech
67433-1-IG-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
The CYP20A1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CYP20A1. This antibody recognizes human, mouse, rat antigen. The CYP20A1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spezifikation
| CYP20A1 | |
| 500 μg/mL | |
| Monoclonal | |
| Mouse | |
| 57404 | |
| Primary |
| 2A9E2 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Human, Rat, Mouse | |
| IgG1 |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.