missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CYSLTR2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16668544
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16668544 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16668544 Supplier Invitrogen™ Supplier No. PA5102072

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CLTR2.

Receptor for cysteinyl leukotrienes. The response is mediated via a G-protein that activates a phosphatidylinositol-calcium second messenger system. Stimulation by BAY u9773, a partial agonist, induces specific contractions of pulmonary veins and might also have an indirect role in the relaxation of the pulmonary vascular endothelium. The rank order of affinities for the leukotrienes is LTC4 = LTD4 >> LTE4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYSLTR2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CYSLTR2
Gene Accession No. Q9NS75
Gene Alias 2300001H05Rik; Cltr2; CysLT(2); Cyslt2; CysLT-2; CYSLT2R; CYSLTR2; cysteinyl leukotriene CysLT2 receptor; cysteinyl leukotriene receptor 2; GPCR21; G-protein coupled receptor GPCR21; G-protein coupled receptor HG57; HG57; hGPCR21; HPN321; KPG_011; PSEC0146; RSBPT32
Gene Symbols CYSLTR2
Host Species Rabbit
Immunogen A synthesized peptide derived from human CYSLTR2(Accession Q9NS75), corresponding to amino acid residues I250-F300.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57105
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.