missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 14/17 Mouse anti-Canine, Feline, Human, Porcine, Clone: EBS-IF-005, Invitrogen™

Product Code. 17165213
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17165213 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17165213 Supplier Invitrogen Supplier No. MA533318

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA5-33318 reacts in immunoblotting specifically with keratin 14 (50kDa). In tissue sections MA5-33318 is positive on basal cells of non-keratinizing stratified epithelia, basal cells and suprabasal cells of the epidermis and gingiva, myoepithelial cells and squamous cell carcinomas.

Cytokeratins(CK) are intermediate filaments of epithelial cells, both in keratinizing tissue (ie. Skin) and non-keratinizing cells (ie. Mesothelial cells). Although not a traditional marker for endothelial cells, cytokeratins have also been found in some microvascular endothelial cells. Atleast 20 different CKs in the molecular range of 40-70 k Da and isoelectric points of 5-805 can be identified using two dimensional gel electrophoresis. Biochemically, most members of the CK family fall into one of two classes, type I (acidic polypeptides) and type II (basic polypeptides). At least one member of the acidic family and one member of the basic family is expressed in all epithelial cells. Monoclonal antibodies to cytokeratin proteins can be useful markers for tumor identification and classification. Belonging to the type A (acidic) subfamily of low molecular weight keratins and existing in combination with keratin 5, keratin 14 distinguishes stratified epithelial cells from simple epithelial cells and is useful in identification of squamous cell carcinomas. It is considered a prognostic marker in breast carcinomas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 14/17
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone EBS-IF-005
Concentration 125 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene KRT17
Gene Accession No. P02533, Q04695
Gene Symbols KRT14, KRT17, LOC100737113
Host Species Mouse
Immunogen Cytokeratin-enriched extract of human ectocervical epithelium
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100685323, 100737113, 101090912, 101091416, 3861, 3872, 490977
Target Species Canine, Feline, Human, Pig
Content And Storage 4° C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.