Learn More
Cytokeratin 3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-91997-0.02ml
This item is not returnable.
View return policy
Description
Cytokeratin 3 Polyclonal antibody specifically detects Cytokeratin 3 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Cytokeratin 3 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| CK3, CK-3, cytokeratin-3, FLJ95909, K3cytokeratin 3, keratin 3, keratin, type II cytoskeletal 3,65 kDa cytokeratin, Keratin-3, Type-II keratin Kb3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 230-390 of human KRT3 (NP_476429.2). WNLLQQQGTSSISGTNNLEPLFENHINYLRSYLDNILGERGRLDSELKNMEDLVEDFKKKYEDEINKRTAAENEFVTLKKDVDSAYMNKVELQAKVDALIDEIDFLRTLYDAELSQMQSHISDTSVVLSMDNNRSLDLDSIIAEVRAQYEDIAQRSKAEAE | |
| 0.02 mL | |
| Cell Biology, Cytoskeleton Markers, Immunology, Neuroscience, Signal Transduction | |
| 3850 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |