missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00€ - 476.00€
Specifications
| Antigen | Cytokeratin 3 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18601061
|
Novus Biologicals
NBP2-91997-0.02ml |
0.02 mL |
190.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18663831
|
Novus Biologicals
NBP2-91997-0.1ml |
0.1 mL |
476.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytokeratin 3 Polyclonal antibody specifically detects Cytokeratin 3 in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Cytokeratin 3 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Immunology, Neuroscience, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 3850 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CK3, CK-3, cytokeratin-3, FLJ95909, K3cytokeratin 3, keratin 3, keratin, type II cytoskeletal 3,65 kDa cytokeratin, Keratin-3, Type-II keratin Kb3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 230-390 of human KRT3 (NP_476429.2). WNLLQQQGTSSISGTNNLEPLFENHINYLRSYLDNILGERGRLDSELKNMEDLVEDFKKKYEDEINKRTAAENEFVTLKKDVDSAYMNKVELQAKVDALIDEIDFLRTLYDAELSQMQSHISDTSVVLSMDNNRSLDLDSIIAEVRAQYEDIAQRSKAEAE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title