Learn More
Epredia™ Lab Vision™ Cytokeratin 8/18 Ab-1, Mouse Monoclonal Antibody
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Cytokeratin 8/18 Ab-1, Mouse Monoclonal Antibody.
Brand: Epredia™ MS-743-S1
Additional Details : Weight : 0.00190kg
Description
Keratin 8 belongs to the type B (basic) subfamily of high molecular weight keratins and exists in combination with cytokeratin 18 (type A; acidic subfamily of low molecular weight keratins).Host Species: Mouse
Clone: 5D3
Isotype: IgG2a
Species Reactivity: Human. Others not known.
Immunogen: Cytokeratins purified from breast cancer MCF-7 cells
Molecular Weight: 52.5 kDa (Cytokeratin 8) and 45 kDa (Cytokeratin 18)
Positive Control: Skin
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Cytokeratin 8/18 Ab-1 | |
Monoclonal | |
Unconjugated | |
Cytokeratins purified from breast cancer MCF-7 cells | |
IVD | |
Primary | |
Concentrated |
Immunohistochemistry (Paraffin) | |
5D3 | |
Mouse | |
500 μL | |
Cytoskeletal Protein | |
Human | |
IgG1 |