missing translation for 'onlineSavingsMsg'
Learn More

D 13223 (Flupirtine Metabolite), TRC

Product Code. 30351907
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30351907 10 mg 10mg
30351017 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30351907 Supplier Toronto Research Chemicals Supplier No. TRCD10006510MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95777-69-8
Molecular Formula C14H16ClFN4O
Molecular Weight (g/mol) 310.75
Synonym D 13223, Flupirtine Metabolite
IUPAC Name N-[2-amino-6-[(4-fluorophenyl)methylamino]pyridin-3-yl]acetamide;hydrochloride
SMILES Cl.CC(=O)Nc1ccc(NCc2ccc(F)cc2)nc1N

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C14H15FN4O.ClH/c1-9(20)18-12-6-7-13(19-14(12)16)17-8-10-2-4-11(15)5-3-10;/h2-7H,8H2,1H3,(H,18,20)(H3,16,17,19);1H
Formula Weight 310.0997
Chemical Name or Material N-(2-Amino-6-(((4-fluorophenyl)methyl)amino)-3-pyridinyl)acetamide Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.