missing translation for 'onlineSavingsMsg'
Learn More

D,L-O-Desmethyl Venlafaxine (100 μg/mL in Methanol), TRC

Product Code. 30407545
Change view
Click to view available options
Quantity:
5 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30407545 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30407545 Supplier Toronto Research Chemicals Supplier No. TRCKIT11955X1ML
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 93413-62-8
Molecular Formula C16 H25 N O2
Quantity 5 x 1 mL
Synonym O-Desmethylvenlafaxine, Phenol, 4-[2-(dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]-, 4-[2-(Dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol, D-VENIZ, DVS 233, Desvenlafaxine, MDD-XR, NEWVEN, O-Desvenlafaxine, Pristiq
InChI Formula InChI=1S/C16H25NO2/c1-17(2)12-15(13-6-8-14(18)9-7-13)16(19)10-4-3-5-11-16/h6-9,15,18-19H,3-5,10-12H2,1-2H3
SMILES CN(C)CC(c1ccc(O)cc1)C2(O)CCCCC2
IUPAC Name 4-[2-(dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol
Molecular Weight (g/mol) 263.38
Formula Weight 263.1885
Concentration or Composition (by Analyte or Components) O-Desmethylvenlafaxine 100 μg/mL
Chemical Name or Material O-Desmethylvenlafaxine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.