missing translation for 'onlineSavingsMsg'
Learn More

D,L-O-Desmethyl Venlafaxine, TRC

Product Code. 17982745
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17982745 5 mg 5mg
17901555 10 mg 10mg
17971765 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17982745 Supplier Toronto Research Chemicals Supplier No. TRCD2965005MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93413-62-8
Molecular Formula C16 H25 N O2
Molecular Weight (g/mol) 263.38
Synonym O-Desmethylvenlafaxine, Phenol, 4-[2-(dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]-, 4-[2-(Dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol, D-VENIZ, DVS 233, Desvenlafaxine, MDD-XR, NEWVEN, O-Desvenlafaxine, Pristiq
IUPAC Name 4-[2-(dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol
SMILES CN(C)CC(c1ccc(O)cc1)C2(O)CCCCC2

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C16H25NO2/c1-17(2)12-15(13-6-8-14(18)9-7-13)16(19)10-4-3-5-11-16/h6-9,15,18-19H,3-5,10-12H2,1-2H3
Formula Weight 263.1885
Percent Purity >95%
Chemical Name or Material O-Desmethylvenlafaxine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.