missing translation for 'onlineSavingsMsg'
Learn More

DDRGK1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

483.00€

Specifications

Antigen DDRGK1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18215023
View Documents
Novus Biologicals
NBP1-59378
100 μL
483.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DDRGK1 Polyclonal specifically detects DDRGK1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

DDRGK1
Polyclonal
Rabbit
Q96HY6
65992
Synthetic peptides corresponding to C20ORF116 The peptide sequence was selected from the middle region of C20ORF116. Peptide sequence EERKRLESQREAEWKKEEERLRLEEEQKEEEERKAREEQAQREHEEYLKL.
Primary
Western Blot
Unconjugated
RUO
C20orf116, chromosome 20 open reading frame 116, DDRGK domain containing 1, DDRGK domain-containing protein 1, dJ1187M17.3, MGC2592
DDRGK1
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.