missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX3 Antibody (3B9E3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-52457
This item is not returnable.
View return policy
Description
DDX3 Monoclonal specifically detects DDX3 in Human, Mouse, Rat, Hamster, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence, Immunocytochemistry, CyTOF-ready.
Specifications
| DDX3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, Immunofluorescence, Immunocytochemistry 1:200-1:1000, CyTOF-ready | |
| DBXCAP-Rf, DDX14, DDX3DEAD/H box-3, DEAD (Asp-Glu-Ala-Asp) box polypeptide 3, X-linked, DEAD box protein 3, X-chromosomal, DEAD box, X isoform, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 3, EC 3.6.1, EC 3.6.4.13, helicase like protein 2, Helicase-like protein 2, HLP2ATP-dependent RNA helicase DDX3X | |
| Mouse | |
| 73.2 kDa | |
| 0.1 mg | |
| Cancer, Tumor Suppressors | |
| 1654 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| 3B9E3 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| DDX3X | |
| Purified recombinant fragment of human DDX3X (AA: 518-661) expressed in E. Coli. | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction