missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX3 Antibody (3B9E3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
196.00€ - 582.00€
Specifications
| Antigen | DDX3 |
|---|---|
| Clone | 3B9E3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, Immunofluorescence, Immunocytochemistry 1:200-1:1000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18127989
|
Novus Biologicals
NBP2-52457 |
0.1 mg |
582.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18631169
|
Novus Biologicals
NBP2-52457-0.025mg |
0.025 mg |
196.00€
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DDX3 Monoclonal specifically detects DDX3 in Human, Mouse, Rat, Hamster, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence, Immunocytochemistry, CyTOF-ready.Specifications
| DDX3 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| Mouse | |
| Cancer, Tumor Suppressors | |
| PBS with 0.05% Sodium Azide | |
| 1654 | |
| Purified recombinant fragment of human DDX3X (AA: 518-661) expressed in E. Coli. | |
| Primary | |
| 73.2 kDa |
| 3B9E3 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, Immunofluorescence, Immunocytochemistry 1:200-1:1000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Monkey | |
| DBXCAP-Rf, DDX14, DDX3DEAD/H box-3, DEAD (Asp-Glu-Ala-Asp) box polypeptide 3, X-linked, DEAD box protein 3, X-chromosomal, DEAD box, X isoform, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 3, EC 3.6.1, EC 3.6.4.13, helicase like protein 2, Helicase-like protein 2, HLP2ATP-dependent RNA helicase DDX3X | |
| DDX3X | |
| IgG2a | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title