missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DDX52 Polyclonal Antibody

Product Code. 16347054
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16347054 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16347054 Supplier Invitrogen™ Supplier No. PA578354

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: Raji Predicted Reactivity: Mouse (84%), Rat (83%), Bovine (85%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The product encoded by this gene is a member of a RNA-binding protein family and possesses nucleolytic activity against cytotoxic lymphocyte (CTL) target cells. It has been suggested that this protein may be involved in the induction of apoptosis as it preferentially recognizes poly(A) homopolymers and induces DNA fragmentation in CTL targets. The major granule-associated species is a 15-kDa protein that is thought to be derived from the carboxyl terminus of the 40-kDa product by proteolytic processing. Alternative splicing resulting in different isoforms of this gene product has been described in the literature.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX52
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.95 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene DDX52
Gene Accession No. Q9Y2R4
Gene Alias 2700029C06Rik; ATP-dependent RNA helicase ROK1-like; ATP-dependent, RNA helicase; DDX52; DEAD (Asp-Glu-Ala-Asp) box polypeptide 52; DEAD box protein 52; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 52; DEAD-box helicase 52; HUSSY19; HUSSY-19; probable ATP-dependent RNA helicase DDX52; Rok1; rROK1L
Gene Symbols DDX52
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human DDX52. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11056
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.