missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DDX52 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17227873 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17227873 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17227873 Supplier Invitrogen™ Supplier No. PA5115107

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DDX52.

The product encoded by this gene is a member of a RNA-binding protein family and possesses nucleolytic activity against cytotoxic lymphocyte (CTL) target cells. It has been suggested that this protein may be involved in the induction of apoptosis as it preferentially recognizes poly(A) homopolymers and induces DNA fragmentation in CTL targets. The major granule-associated species is a 15-kDa protein that is thought to be derived from the carboxyl terminus of the 40-kDa product by proteolytic processing. Alternative splicing resulting in different isoforms of this gene product has been described in the literature.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDX52
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene DDX52
Gene Accession No. Q9Y2R4
Gene Alias 2700029C06Rik; ATP-dependent RNA helicase ROK1-like; ATP-dependent, RNA helicase; DDX52; DEAD (Asp-Glu-Ala-Asp) box polypeptide 52; DEAD box protein 52; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 52; DEAD-box helicase 52; HUSSY19; HUSSY-19; probable ATP-dependent RNA helicase DDX52; Rok1; rROK1L
Gene Symbols DDX52
Host Species Rabbit
Immunogen A synthesized peptide derived from human DDX52(Accession Q9Y2R4), corresponding to amino acid residues K553-S599.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11056
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.