missing translation for 'onlineSavingsMsg'
Learn More

DEC-205/CD205 Antibody [HRP], Novus Biologicals™

Product Code. 30760012 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30760012 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30760012 Supplier Novus Biologicals Supplier No. AF5975H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

DEC-205/CD205 Polyclonal antibody specifically detects DEC-205/CD205 in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DEC-205/CD205
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias CD205, CLEC13B, C-type lectin domain family 13 member B, DEC-205, GP200-MR6, Ly-75, Lymphocyte antigen 75
Host Species Goat
Immunogen E. coli-derived recombinant mouse DEC-205/CD205, Cys216-Pro503, Accession # Q60767
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Dendritic Cell Markers, Ovarian Carcinoma Cell Markers, Renal Cell Carcinoma Cell Markers, Vision
Primary or Secondary Primary
Gene ID (Entrez) 4065
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.