missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ alpha-Defensin 3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16669574
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16669574 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16669574 Supplier Invitrogen™ Supplier No. PA5103123

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Defensin alpha3.

Defensins are a family of antimicrobial and cytotoxic peptides thought to be involved in host defense. They are abundant in the granules of neutrophils and also found in the epithelia of mucosal surfaces such as those of the intestine, respiratory tract, urinary tract, and vagina. Members of the defensin family are highly similar in protein sequence and distinguished by a conserved cysteine motif. The protein encoded by this gene, defensin, alpha 3, is found in the microbicidal granules of neutrophils and likely plays a role in phagocyte-mediated host defense. Several alpha defensin genes are clustered on chromosome 8. This gene differs from defensin, alpha 1 by only one amino acid.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha-Defensin 3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DEFA3
Gene Accession No. P28310, P59666
Gene Alias alpha-defensin 1; Alpha-defensin 3; cryptdin-1; DEF1; DEF3; Defa1; DEFA1B; DEFA2; Defa27; Defa3; Defcr; Defcr1; Defcr3; Defcr-3; defensin 3, neutrophil-specific; defensin alpha 1; defensin alpha 3; defensin related cryptdin peptide 1; defensin, alpha 1; defensin, alpha 1, myeloid-related sequence; defensin, alpha 2; Defensin, alpha 3; defensin, alpha 3, neutrophil-specific; defensin, alpha, 27; defensin, alpha, 3; defensin-1; defensin-related cryptdin peptide; Defensin-related cryptdin-3; HNP-1; HNP-2; HNP3; HNP-3; HP 1-56; HP 3-56; HP1; HP-1; HP-2; HP3; HP-3; MRS; myeloid-related sequence; neutrophil defensin 1; Neutrophil defensin 2; Neutrophil defensin 3; neutrophil peptide 3
Gene Symbols DEFA3
Host Species Rabbit
Immunogen A synthesized peptide derived from human DEFA3(Accession P59666), corresponding to amino acid residues I36-Q86.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13237, 1668
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.