missing translation for 'onlineSavingsMsg'
Learn More

Delta 1 Tubulin Antibody, Novus Biologicals™

Product Code. 18007034 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.2 mL
Unit Size:
0.2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18007034 0.2 mL 0.2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18007034 Supplier Novus Biologicals Supplier No. NB10057397

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Delta 1 Tubulin Polyclonal specifically detects Delta 1 Tubulin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Delta 1 Tubulin
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500- 1:2 000, ELISA, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation Antiserum with No Preservative
Gene Accession No. Q9UJT1
Gene Alias delta-tubulin, TUBDFLJ12709, tubulin delta chain, tubulin, delta 1
Gene Symbols TUBD1
Host Species Rabbit
Immunogen Synthetic peptides derived from Cter domain of human tubulin
Purification Method Unpurified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51174
Test Specificity Tubulin Delta 2
Reconstitution Reconstitute with 0.2 ml distilled water.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.