missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dengue Virus Capsid Protein Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12745-50ul
This item is not returnable.
View return policy
Description
Dengue Virus Capsid Protein Polyclonal antibody specifically detects Dengue Virus Capsid Protein in Virus samples. It is validated for Western Blot
Specifications
| Dengue Virus Capsid Protein | |
| Polyclonal | |
| Western Blot 1:1000-1:5000 | |
| polyprotein gene | |
| Produced in rabbits immunized with purified, recombinant Dengue Virus Capsid Protein (Accession#: AHF52833.1; Met1-Arg100). | |
| 50 μg | |
| Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| 1494449 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction