missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dengue Virus Capsid Protein Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
299.00€ - 507.00€
Specifications
| Antigen | Dengue Virus Capsid Protein |
|---|---|
| Dilution | Western Blot 1:1000-1:5000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18692449
|
Novus Biologicals
NBP3-12745-100ul |
100 μg |
507.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18620677
|
Novus Biologicals
NBP3-12745-50ul |
50 μg |
299.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dengue Virus Capsid Protein Polyclonal antibody specifically detects Dengue Virus Capsid Protein in Virus samples. It is validated for Western BlotSpecifications
| Dengue Virus Capsid Protein | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| 0.2 um filtered solution in PBS | |
| 1494449 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:1000-1:5000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Virus | |
| polyprotein gene | |
| Produced in rabbits immunized with purified, recombinant Dengue Virus Capsid Protein (Accession#: AHF52833.1; Met1-Arg100). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title