Learn More
DEPDC6/Deptor Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Shop All Proteintech ProductsDescription
The DEPDC6/Deptor antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human DEPDC6/Deptor. This antibody recognizes human, mouse, rat antigen. The DEPDC6/Deptor antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
| Antigen | DEPDC6/Deptor |
| Applications | Immunofluorescence |
| Concentration | 1000 μg/mL |
| Conjugate | CoraLite Plus 488 |
| Gene Symbols | DEP domain containing 6, DEP.6, DEPDC6, Deptor, DKFZp564B1778, FLJ12341, FLJ12428, FLJ13854 |
| Host Species | Rabbit |
| Quantity | 20 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 64798 |
| Target Species | Human, Mouse, Rat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.